ENA405194400 Enamine

3-(2-chloro-6-fluorophenyl)piperazin-2-one

  • MDL number MFCD16715584
  • Cas Number : 1248210-76-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175...177
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133876

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133876
Availability details for
Quantity
  • Availability
  • Add to Cart