ENA405194418 Enamine

N-(2,4-dimethylphenyl)-3-hydroxynaphthalene-2-carboxamide

  • MDL number MFCD00021633
  • Cas Number : 92-75-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 226...228
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133963

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133963
Availability details for
Quantity
  • Availability
  • Add to Cart