ENA405194420 Enamine

2-chloro-5-[(cyclopropylmethoxy)methyl]pyridine

  • MDL number MFCD11136687
  • Cas Number : 1019546-38-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133984

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133984
Availability details for
Quantity
  • Availability
  • Add to Cart