ENA405194424 Enamine

2-Chloro-N-(2-chloro-phenyl)-acetamide

  • MDL number MFCD00032560
  • Cas Number : 3289-76-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78...80
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01510

Safety Information

msds https://www.enaminestore.com/catalog/EN300-01510
Availability details for
Quantity
  • Availability
  • Add to Cart