ENA405194428 Enamine

[4-(dimethylamino)-3-nitrophenyl]methanol

  • MDL number MFCD11191404
  • Cas Number : 1039882-00-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134033

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134033
Availability details for
Quantity
  • Availability
  • Add to Cart