ENA405194431 Enamine

3-(4-methylpyrrolidin-2-yl)pyridine

  • MDL number MFCD05215065
  • Cas Number : 603090-13-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134036

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134036
Availability details for
Quantity
  • Availability
  • Add to Cart