ENA405194433 Enamine

3-(3-fluorophenyl)prop-2-enenitrile

  • MDL number MFCD16083018
  • Cas Number : 1383934-14-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 52...54
Purity 95.00
Flash Point 0
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134038

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134038
Availability details for
Quantity
  • Availability
  • Add to Cart