ENA405194443 Enamine

1-tert-butyl-2-(4-chloro-3-fluorophenyl)-5-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD25970265
  • Cas Number : 1408761-55-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 249...251
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128797

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128797
Availability details for
Quantity
  • Availability
  • Add to Cart