ENA405194445 Enamine

2-benzyl-octahydro-1H-isoindol-4-amine

  • MDL number MFCD10758104
  • Cas Number : 217464-22-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128806

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128806
Availability details for
Quantity
  • Availability
  • Add to Cart