ENA405194451 Enamine

8-chloro-3,4-dihydro-2H-1-benzopyran-4-ol

  • MDL number MFCD11899781
  • Cas Number : 1176104-70-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134063

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134063
Availability details for
Quantity
  • Availability
  • Add to Cart