ENA405194453 Enamine

methyl 6-[(3-hydroxyphenyl)formamido]hexanoate

  • MDL number MFCD12487733
  • Cas Number : 1156079-14-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134065

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134065
Availability details for
Quantity
  • Availability
  • Add to Cart