ENA405194455 Enamine

methyl 2-[4-(methylamino)phenoxy]acetate hydrochloride

  • MDL number MFCD25970408
  • Cas Number : 1461714-23-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158...160
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134068

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134068
Availability details for
Quantity
  • Availability
  • Add to Cart