ENA405194456 Enamine

1-(oxan-4-yl)guanidine hydrochloride

  • MDL number MFCD25970409
  • Cas Number : 1447606-48-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 196...198
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134069

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-134069
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart