ENA405194462 Enamine

1-methyl-1H-indazole-3-carboxamide

  • MDL number MFCD21604247
  • Cas Number : 129137-93-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166...168
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134079

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134079
Availability details for
Quantity
  • Availability
  • Add to Cart