ENA405194463 Enamine

methyl 1-methyl-1H-indazole-3-carboxylate

  • MDL number MFCD03840643
  • Cas Number : 109216-60-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-134081

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134081
Availability details for
Quantity
  • Availability
  • Add to Cart