ENA405194466 Enamine

3-(3-phenyl-1H-1,2,4-triazol-5-yl)piperidine dihydrochloride

  • MDL number MFCD25970413
  • Cas Number : 1461713-40-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134105

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134105
Availability details for
Quantity
  • Availability
  • Add to Cart