ENA405194474 Enamine

2-(1,3-thiazol-2-yl)phenol

  • MDL number MFCD14529356
  • Cas Number : 83053-38-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 50-52
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134117

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134117
Availability details for
Quantity
  • Availability
  • Add to Cart