ENA405194479 Enamine

1-(3,4-difluorophenyl)-2-(propan-2-yloxy)ethan-1-ol

  • MDL number MFCD21953130
  • Cas Number : 1461713-71-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-134129

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134129
Availability details for
Quantity
  • Availability
  • Add to Cart