ENA405194481 Enamine

4-bromo-1-(difluoromethyl)-2-nitrobenzene

  • MDL number MFCD18391144
  • Cas Number : 1261442-15-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128938

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128938
Availability details for
Quantity
  • Availability
  • Add to Cart