ENA405194483 Enamine

5-bromo-2-chloro-N-methylpyrimidin-4-amine

  • MDL number MFCD09863053
  • Cas Number : 205672-24-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149...151
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134133

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134133
Availability details for
Quantity
  • Availability
  • Add to Cart