ENA405194486 Enamine

3-methyl-1-(pyridin-4-yl)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD12134545
  • Cas Number : 1153043-06-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150...152
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134140

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134140
Availability details for
Quantity
  • Availability
  • Add to Cart