ENA405194487 Enamine

6-fluoro-8-nitro-3,4-dihydro-2H-1-benzopyran-4-one

  • MDL number MFCD11518437
  • Cas Number : 258528-35-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108...110
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134141

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134141
Availability details for
Quantity
  • Availability
  • Add to Cart