ENA405194488 Enamine

4-aminobenzene-1-sulfonyl fluoride

  • MDL number MFCD00007883
  • Cas Number : 98-62-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 67...69
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134143

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-134143
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart