ENA405194491 Enamine

methyl 3-[(2-methoxy-2-oxoethyl)(propan-2-yl)amino]propanoate

  • MDL number MFCD16160595
  • Cas Number : 1249253-89-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129132

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129132
Availability details for
Quantity
  • Availability
  • Add to Cart