ENA405194501 Enamine

[1-(4-aminophenyl)-1H-1,2,3-triazol-4-yl]methanol

  • MDL number MFCD11844966
  • Cas Number : 1026133-62-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-134218

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134218
Availability details for
Quantity
  • Availability
  • Add to Cart