ENA405194512 Enamine

1-methyl-4,5-dihydro-1H-1,2,4-triazole-5-thione

  • MDL number MFCD00128149
  • Cas Number : 32362-75-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 186...188
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134247

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134247
Availability details for
Quantity
  • Availability
  • Add to Cart