ENA405194516 Enamine

5-[4-(trifluoromethyl)phenyl]-1,3-oxazolidin-2-one

  • MDL number MFCD21266605
  • Cas Number : 1461715-50-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112...114
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134252

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134252
Availability details for
Quantity
  • Availability
  • Add to Cart