ENA405194517 Enamine

(2R)-1-(1,3-dioxaindan-5-yl)propan-2-ol

  • MDL number MFCD11111319
  • Cas Number : 521097-97-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134253

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134253
Availability details for
Quantity
  • Availability
  • Add to Cart