ENA405194519 Enamine

ethyl 3-amino-2-methylbutanoate

  • MDL number MFCD19205384
  • Cas Number : 4946-35-4
  • Molecular Weight : 145.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 3-amino-2-methylbutanoate
Inchi Code 1S/C7H15NO2/c1-4-10-7(9)5(2)6(3)8/h5-6H,4,8H2,1-3H3
Inchi Key FMLDWVZXTNBACN-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-134256

Safety Information

signalword Danger
pcodes P210,P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P310,P312,P332+P313,P362,P370+P378,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-134256
pictograms GHS05,GHS07
hstatements H227,H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart