ENA405194521 Enamine

1-(2H-1,3-benzodioxol-5-yl)propan-2-ol

  • MDL number MFCD16251522
  • Cas Number : 6974-61-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134264

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134264
Availability details for
Quantity
  • Availability
  • Add to Cart