ENA405194522 Enamine

methyl 3-{[(tert-butoxy)carbonyl](methyl)amino}-2-methylpropanoate

  • MDL number MFCD16057136
  • Cas Number : 1248914-26-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134265

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134265
Availability details for
Quantity
  • Availability
  • Add to Cart