ENA405194524 Enamine

(1S)-1-cycloheptylethan-1-ol

  • MDL number MFCD21197604
  • Cas Number : 1461689-19-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134267

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134267
Availability details for
Quantity
  • Availability
  • Add to Cart