ENA405194528 Enamine

1-azatricyclo[6.3.1.0^{4,12}]dodeca-4(12),5,7-trien-2-one

  • MDL number MFCD08693146
  • Cas Number : 16078-37-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88...90
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134280

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134280
Availability details for
Quantity
  • Availability
  • Add to Cart