ENA405194532 Enamine

6,7-dibromo-4-hydroxy-1H,2H,3H,4H-pyrrolo[1,2-a]pyrazin-1-one

  • MDL number MFCD25970428
  • Cas Number : 219783-00-3
  • Molecular Weight : 309.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-134296

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134296
Availability details for
Quantity
  • Availability
  • Add to Cart