ENA405194536 Enamine

2-Chloro-N-(3,5-dimethoxy-phenyl)-acetamide

  • MDL number MFCD00751234
  • Cas Number : 66932-96-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01701

Safety Information

msds https://www.enaminestore.com/catalog/EN300-01701
Availability details for
Quantity
  • Availability
  • Add to Cart