ENA405194538 Enamine

methyl 1-(pyridin-3-yl)-1H-1,2,3-triazole-4-carboxylate

  • MDL number MFCD25970431
  • Cas Number : 1012081-63-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169...171
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134346

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134346
Availability details for
Quantity
  • Availability
  • Add to Cart