ENA405194544 Enamine

3-(2,4-difluorophenyl)prop-2-enal

  • MDL number MFCD07437912
  • Cas Number : 883107-63-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63...65
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134363

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134363
Availability details for
Quantity
  • Availability
  • Add to Cart