ENA405194556 Enamine

2-[(4-chlorophenyl)sulfanyl]-4-methylbenzonitrile

  • MDL number MFCD18979115
  • Cas Number : 1405886-75-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134379

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134379
Availability details for
Quantity
  • Availability
  • Add to Cart