ENA405194557 Enamine

4-(2-methylphenyl)piperidine hydrochloride

  • MDL number MFCD02178908
  • Cas Number : 82212-02-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 287...289
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134382

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134382
Availability details for
Quantity
  • Availability
  • Add to Cart