ENA405194585 Enamine

methyl 3-[(3-aminopyridin-2-yl)sulfanyl]propanoate

  • MDL number MFCD12092176
  • Cas Number : 111042-83-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134502

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134502
Availability details for
Quantity
  • Availability
  • Add to Cart