ENA405194590 Enamine

5-(1-chloroethyl)-3-(methoxymethyl)-1,2,4-oxadiazole

  • MDL number MFCD14690898
  • Cas Number : 1250158-52-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134508

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134508
Availability details for
Quantity
  • Availability
  • Add to Cart