ENA405194595 Enamine

5-methoxy-7-methyl-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD11007874
  • Cas Number : 343774-48-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134514

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134514
Availability details for
Quantity
  • Availability
  • Add to Cart