ENA405194601 Enamine

1-bromo-2-methoxy-2-methylpropane

  • MDL number MFCD08063237
  • Cas Number : 19752-21-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134530

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-134530
pictograms GHS06
Availability details for
Quantity
  • Availability
  • Add to Cart