ENA405194602 Enamine

4-propoxybenzene-1-sulfonamide

  • MDL number MFCD00460574
  • Cas Number : 1135-01-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119...121
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134531

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134531
Availability details for
Quantity
  • Availability
  • Add to Cart