ENA405194608 Enamine

5-bromo-N-(2-methoxyphenyl)pyrimidin-2-amine

  • MDL number MFCD19019798
  • Cas Number : 1410670-75-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134553

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134553
Availability details for
Quantity
  • Availability
  • Add to Cart