ENA405194611 Enamine

3-(3-fluoro-4-methoxyphenyl)-2-methylprop-2-enoic acid

  • MDL number MFCD06207001
  • Cas Number : 32004-62-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 168...170
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130053

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130053
Availability details for
Quantity
  • Availability
  • Add to Cart