ENA405194612 Enamine

3-(3-fluoro-4-methoxyphenyl)-2-methylpropanoic acid

  • MDL number MFCD12794483
  • Cas Number : 50703-56-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130054

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130054
Availability details for
Quantity
  • Availability
  • Add to Cart