ENA405194616 Enamine

3-(3-fluorophenoxy)pyrrolidine hydrochloride

  • MDL number MFCD09879239
  • Cas Number : 1185119-76-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 106...108
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134564

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-134564
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart