ENA405194628 Enamine

1-benzylcyclopentan-1-amine hydrochloride

  • MDL number MFCD01735698
  • Cas Number : 20937-44-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206...208
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134620

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134620
Availability details for
Quantity
  • Availability
  • Add to Cart