ENA405194634 Enamine

1-[5-(4-chloro-2-fluorophenyl)thiophen-2-yl]ethan-1-one

  • MDL number MFCD11206992
  • Cas Number : 1094282-07-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134636

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134636
Availability details for
Quantity
  • Availability
  • Add to Cart